missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Heathrow Scientific Droplet™ Phlebotomy/Sample Collection Tray Kit, Blue/Green
Description
- Multiple compartments keep a variety of supplies neatly at hand and organized
- Built in side pockets offer additional storage space
- Handle includes soft rubber grip to reduce hand and finger strain
- Each tray includes either a 13 or 16 mm OneRack™ tube rack and two disposable polystyrene inserts
- Ratcheting Handle folds out of the way for easy access, even over tall sharps containers
- Includes side pockets and supply drawer
Specifications
Specifications
| Product Type | Phlebotomy Tray |
| Includes | 16 mm Tube Rack Insert, 2 Disposable Inserts |
| Dimensions (L x W x H) | 16.8 x 10.4 x 6.7 in. (42.9 x 26.5 x 17.2 cm) |
| Height (English) | 11 in. |
| Material | ABS |