missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Drilled Brass Ball

Catalog No. S24486
Change view
Click to view available options
1 in.; With 2.5mm drilled hole
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
S24486 1 in.; With 2.5mm drilled hole
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S24486 Supplier Eisco™ Supplier No. BLDBL25

For pendulum demonstrations in lab

  • With 2.5mm drilled hole
  • Drilled for lab demonstrations

Specifications

Product Type Brass Ball
For Use With (Application) Pendulum demonstration
Includes With 2.5mm drilled hole
Height (English) 1 in.