missing translation for 'onlineSavingsMsg'
Learn More

DRIFIRE™ Fortrex™ Flight Suit Pants
GREENER_CHOICE

Catalog No. 17000255
Change view
Click to view available options
Size (English) Waist:
27 to 31 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size (English) Waist
17000255 27 to 31 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17000255 Supplier DRIFIRE™ Supplier No. 27189

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Unique design enables reduced binding, increased range of motion and reduced snag hazards

  • Made from DRIFIRE's proprietary FORTREX fabric
  • Permanent, inherent flame resistance that will not wash or wear out
  • Reinforced pant button to prevent F.O.D.
  • Flexible 2-piece design increases air flow
  • Antimicrobial protection to resist odor and improve overall hygiene
  • Wicking and fast-drying
  • Suitable for both wet and dry conditions
  • Good tensile and tear strengths
  • Meets or exceeds Air Force 2-piece Flight Suit and Army A2CU requirements
  • Available in multicam or tiger stripe prints

Easily accessible storage pockets and velcroL:

  • Chest pockets made for easy access, even if worn under body armor
  • Upper arm pockets with dual access compartment and improved pen pocket design
  • Velcro and zipper closures on pant pockets gives added security and adjustable velcro waist tabs for customized fit
  • Velcro cuffs on wrists and ankles offer adjustable fit and minimize rotor distortion

US Army, US Coast Guard, Air Force, US Law Enforcement/Defense Contractors & US SOCOM

Specifications

Size Small
Product Type Flame-resistant Flight Suit Pants
Material Fortrex Fabric
Color Multi-cam
Closure Type Hook and Loop Seal, Zipper
Resistant To Flame
Certifications/Compliance A2CU
Size (English) Waist 27 to 31 in.
Size (Metric) Waist 68.5 to 78.7 cm
For Use With (Application) US army, US coast guard, air force, US law enforcement/defense contractors and US SOCOM