missing translation for 'onlineSavingsMsg'
Learn More

Drierite™ Indicating Absorbents, W.A. Hammond

Catalog No. p-2682741
Change view
Click to view available options
Quantity:
1 lb.
5 lb.
Mesh Size:
10 to 20
4
8
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Mesh Size
075784A 1 lb. 10 to 20
075783A 1 lb. 8
075784B 5 lb. 10 to 20
23116681 5 lb. 4
075783B 5 lb. 8
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 075784A Supplier W A Hammond Drierite Co Supplier No. HDC24001
Only null left

Efficient desiccant giving constant visual indication of the progress of absorption and exhaustion. W A Hammond Drierite™ Drying Dessicants dries organic liquids to low ppm levels and is commonly used in desiccators, desiccator cabinets, in-line air and gas dryers.

  • Efficient desiccant giving constant visual indication of the progress of absorption and exhaustion
  • Turns from blue to pink when exhausted
  • Dries air and gases to lower residual moisture levels (-100°F dew point)
  • Commonly used in desiccators, desiccator cabinets, in-line air and gas drying columns, absorption bulbs, U-tubes, glove boxes , sealed chambers and wherever an anhydrous environment is required
  • Regeneration by heating granules on tray in single layer at 210-220°C for 1 hour

Specifications

Chemical Name or Material Calcium Sulfate with Cobalt Chloride Indicator
CAS 7646-79-9
Material Calcium Sulfate with Cobalt Chloride
Quantity 1 lb.
Molecular Formula 98% CaSO4 with <2% CoCL2
Mesh Size 10 to 20
Packaging Glass Bottle
Synonym Abosrbent; Desiccant; Indicating