missing translation for 'onlineSavingsMsg'
Learn More

DR Instruments DR-Uniforms™ Sanforized 100% Cotton Unisex Lab Coat

Catalog No. S72146
Change view
Click to view available options
Size:
Medium
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
S72146 Medium
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S72146 Supplier DR Instruments Supplier No. DR2012M
Only null left

DR-Uniforms™ Sanforized 100% Cotton Unisex Lab Coats are tailored from fine quality materials for hospital and lab use. Feature three patch pockets and two convenient side slits to access pants pockets.

  • Unisex Lab Coats
  • 100% Cotton - Soft and light weight Twill Fabric
  • Senforized to control shrinkage to the minimum.
  • Mercerized to increases fabric luster, strength, shine, smoothness and higher quality silky appearance.
  • Moisture absorbent.
  • Ideal for chemistry students and doctors alike.
  • Sizes available: XS through 5XL
  • Full-length (42-inch),
  • 3 patch pockets
  • 2 side slits
  • Side slits for access pant pockets

Specifications

Product Type General Purpose Lab Coat
Size Medium
Gender Unisex
Material Cotton
Color White
Closure Type Buttons
Disposable Reusable
Sterility Non-sterile
Style Long
No. of Pockets 3
Length (English) 42 in.
Length (Metric) 106.7 cm
Show More Show Less