Learn More
DR Instruments Scalpel with Safety Lock and #22 Blade
Catalog No. S07898
Change view
Click to view available options
Size:
# 22
Catalog No. Size
S07898 # 22
1 options
Catalog No. S07898 Supplier DR Instruments Supplier No. EPD21
Safety lock scalpel offers secure blade mount and easy replacement of blades. Holds #22, #21, and #23 blades.
  • Stainless steel made scalpel handel with screw lock for blade
  • Comes with one blade, No. 22 (Installed)
  • Non-Slip grip with ridges
Specifications
Blade Material Chrome-plated Steel
Product Type Scalpel
Size # 22
For Use With (Equipment) #22, #21, and #23 Blades
Includes Screw Lock