missing translation for 'onlineSavingsMsg'
Learn More

DR Instruments Iris Scissors, 4.5 in., Stainless Steel

Catalog No. S07897
Change view
Click to view available options
Tip Style:
Curved Fine Points
Fine Point
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Tip Style
S07897 Curved Fine Points
S08088 Fine Point
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options

California Proposition 65 WARNING: Cancer and Reproductive Harm

Catalog No. S07897 Supplier DR Instruments Supplier No. 10

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

California Proposition 65 WARNING: Cancer and Reproductive Harm

Iris Scissors feature precision ground blades for effortless cutting. Ideal for delicate dissections.

  • For fine dissection work
  • Surgical stainless steel construction
  • Available in sand finish or mirror polish finish
  • Length: 11.4 cm (4.5 in.)

Specifications

Length (English) 4.5 in.
Length (Metric) 11.4 cm
Material Stainless Steel
For Use With (Application) Dissection
Tip Style Curved Fine Points