missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DR Instruments Flexible Probe with Eyelet, 6 in., Stainless Steel
Flexible Probe with Eyelet used for all dissecting applications.
Supplier: DR Instruments 35FLX
Description
- One end features an eyelet and the other is tapered with an olive tip
- Crafted with Stainless Steel
- Length: 15.24 cm (6 in.)
Specifications
| Flexible Probe | |
| Stainless Steel | |
| 6 in. | |
| 15.24 cm |