missing translation for 'onlineSavingsMsg'
Learn More

DR Instruments Dissecting Forceps with Curved Medium Points, Guide Pin, 4.5 in.

Catalog No. S08097
Change view
Click to view available options
Tip Style:
Curved Medium Points
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Tip Style
S08097 Curved Medium Points
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S08097 Supplier DR Instruments Supplier No. 14

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Curved tips for flexibility to access hard-to-reach areas during dissection.

  • Crafted with surgical stainless-steel
  • Guide pin for matching of points
  • Ridges for easy grip
  • Length: 11.43 cm (4.5 in.)

Specifications

Product Type Forceps
Material Stainless Steel
Includes Guide Pin
Tip Style Curved Medium Points
Length (English) 4.5 in.
Length (Metric) 11.4 cm