missing translation for 'onlineSavingsMsg'
Learn More

Dräger™ X-plore™ Ammonia/Methylamine (AM/MA) Cartridge

For protection against Ammonia and Methylamine gases

$20.19 - $20.19

Specifications

Certifications/Compliance NIOSH
Color Blue
Material ABS Plastic
Product Type Cartridge
For Use With (Equipment) Mask
View More Specs

Products 1
Catalog Number Mfr. No. Cartridge Type Price Quantity  
Catalog Number Mfr. No. Cartridge Type Price Quantity  
17-985-670
View Documents
Dräger™
6738027
Gas/Vapor Cartridge
Each for $20.19
Only null left
 
Description

Description

Fits Dräger X-plore 3300/3500 half masks and Dräger X-plore 5500 full face masks

  • Strong blue ABS plastic housing
  • Safe, quick and easy bayonet connection with clear guidance bringing the filter/cartridge securely into place
  • Both filters are inserted in a top-down position
  • Option to upgrade a gas cartridge to a combination cartridge by simply adding a pad retrainer and a N 95 or R 95 pad
  • Shelflife (from date of production): gas and combination cartridges 6 years, pads 3 years

  • To convert the AM/MA cartridge to a combination cartridge, you will need two filter pad retainers (p/n: 6738038) and two N95 Prefilters (p/n: 6738038).

Order Info

Packaged in pairs for optimum protection of the unused filter. Instructions included with each pair.

Certifications

NIOSH 42 CFR 84

Specifications

Specifications

NIOSH
ABS Plastic
Mask
Blue
Cartridge
1 Ea.
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'