missing translation for 'onlineSavingsMsg'
Learn More

Dräger™ X-plore™ 5500 PC Full Face Mask

Catalog No. 17985652
Change view
Click to view available options
Suspension Type:
4-point
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Suspension Type
17985652 4-point
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17985652 Supplier Dräger™ Supplier No. R55270

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The Dräger™ X-plore™ 5500 EPDM full face mask is the right solution for environments that require increased respiratory protection and a clear field of vision

The double-layer face seal with triple sealing action ensures reliable protection and a secure, leak-free seal regardless of your face shape

 

  • Double-layer face seal with triple sealing action
  • Bayonet connection for easy and secure filter attachment. Both filters are inserted in a top-down direction
  • The swept-back low profile design ensures an optimal field of vision and even weight distribution with superb freedom of movement
  • Impact-proof polycarbonate material (with plastic frame) lens provides excellent, wide field of vision and extreme chemical, thermal, and mechanical resistance
  • 5-point harness that allows quick and easy donning without hair entanglement
  • Three sizes in one, fits most face and head sizes
  • EPDM mask body with high ageing resistance
  • Two side-positioned bayonet connections for use with the Dräger™ X-plore™ Twinfilter cartridge series
Note: filters are not included; must be ordered separately.

 

Certifications

CE Marked

Compliance

EN136 Cl.2 and NIOSH

Specifications

Certifications/Compliance NIOSH, CE, EN136 Cl.2
Material EPDM, Polycarbonate Lens
Product Type Full Face Mask
Format Universal
For Use With (Application) Right solution for environments that require increased respiratory protection and a clear field of vision
Suspension Type 4-point