Electron Microscopy Sciences DPX Mountant for Microscopy, 100ml
Catalog No. 50980369
DPX Mountant for Microscopy is a mixture of Distyrene, a plasticizer, and xylene. It is a colorless synthetic resin mounting media which replaces Xylene-Balsam. It preserves the stain and dries quickly. It comes in the amount of 100ml.
Catalog No.
50980369
Supplier
Electron Microscopy SciencesSupplier No.
13510
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
DPX Mountant for Microscopy is a mixture of Distyrene, a plasticizer, and xylene. It is a colorless synthetic resin mounting media which replaces Xylene-Balsam. It preserves the stain and dries quickly. It comes in the amount of 100ml.