missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals DPDPE

Catalog No. AAJ66293LB0
Change view
Click to view available options
Quantity:
1 mg
5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ66293LB0 5 mg
AAJ66293MCR 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ66293LB0 Supplier Thermo Scientific Chemicals Supplier No. J66293LB0

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For cellular and molecular biology applications

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Purification Method HPLC
Common Name [D-Pen2,5] Enkephalin hydrate
Content And Storage -30°C to -10°C
Format Solid
Molecular Weight (g/mol) 645.79
Product Type DPDPE
Quantity 5 mg
Regulatory Status RUO
Sequence Tyr-D-Pen-Gly-Phe-D-Pen (Disulfide bridge: D-Pen2-D-Pen5)

RUO – Research Use Only