missing translation for 'onlineSavingsMsg'
Learn More

Lovibond™ DPD No. 3 HR EVO Tablets

The DPD No. 3 EVO Tablet is used in combination with the DPD No. 1 Tablet to measure high ranges (up to 10 mg/L) of Free and Total Chlorine in a water sample.

$64.37 - $269.43

Specifications

Analytes Chlorine
Format Tablet
For Use With (Application) Water Quality Analysis
For Use With (Equipment) Lovibond™ Colorimeters and Spectrophotometers
Sample Size 10 mL
View More Specs

Products 3
Catalog Number Mfr. No. No. of Tests Price Quantity  
Catalog Number Mfr. No. No. of Tests Price Quantity  
01671058
View Documents
Lovibond™
511920BT
100 Tests N/A
Only null left
 
01671059
View Documents
Lovibond™
511921BT
250 Tests N/A
Only null left
 
01671060
View Documents
Lovibond™
511922BT
500 Tests N/A
Only null left
 
Description

Description

With over 50 years of manufacturing water quality reagents, Lovibond™ knows choosing the right test for your process or application is critical for accurate results. Each tablet is protected from humidity, heat and other environmental interferences, as pharmaceutical-grade aluminum blister packaging individually seals up to 10 tablets on an individual strip

  • Easy to dispense blister pack makes dispensing tablets safer
  • Shelf-stable, analytical solution for testing water quality parameters
  • Accurate and precise dosing
  • Long shelf-life
Specifications

Specifications

Chlorine
Water Quality Analysis
10 mL
2 min
Tablet
Lovibond™ Colorimeters and Spectrophotometers
DPD
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'