missing translation for 'onlineSavingsMsg'
Learn More

Lovibond™ DPD No. 1 EVO and No. 3 EVO Combination Pack

Catalog No. 01671065
Change view
Click to view available options
No. of Tests:
100 Tests
250 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Tests
01671065 250 Tests
01671064 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01671065 Supplier Lovibond™ Supplier No. 517932BT

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This combination pack delivers everything needed to perform tests for Free and Total Chlorine in a single water sample.

With over 50 years of manufacturing water quality reagents, Lovibond™ knows choosing the right test for your process or application is critical for accurate results. Each tablet is protected from humidity, heat and other environmental interferences, as pharmaceutical-grade aluminum blister packaging individually seals up to 10 tablets on an individual strip.

  • Easy to dispense blister pack makes dispensing tablets safer
  • Shelf-stable, analytical solution for testing water quality parameters
  • Accurate and precise dosing
  • Long shelf-life

Specifications

Analytes Chlorine
Format Tablet
For Use With (Application) Water Quality Analysis
For Use With (Equipment) Lovibond™ Colorimeters and Spectrophotometers
Includes 250 DPD No. 1 and 250 DPD No. 3 EVO Tablets
No. of Tests 250 Tests
Sample Size 10 mL
Test Method DPD
Test Time 2 min
Type Reagent Tablet