missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LaMotte™ DPD Free Chlorine Reagent Refills
The liquid alternative to DPD tablet refills
Supplier: LaMotte™ P6741CNG
Alert:
Keep out of reach of children. May irritate eyes and skin. Harmful if swallowed.
Description
- DPD £1A and DPD £1B test Free Available Chlorine (Cl)
- DPD £3 is added to the reacted sample to measure Total Cl
- Comes in 30mL and 60mL sizes
DPD = N, N-Diethyl-1, 4-phenylene-diamine sulfate
Use with existing LaMotte chlorine comparators or colorimeters. For each test, all three reagents are added to the same sample. Ideal for testing drinking water, food and beverage, pool and spa, industrial water boiling and cooling, laundry and sanitation, water and wastewater.
Chemical Identifiers
| Measurement of total chlorine, provides quicker results and clearer colors than tablets | |
| LaMotte™ colorimeters and comparators | |
| Each |
Specifications
| 30 mL | |
| LaMotte™ colorimeters and comparators | |
| DPD 1B Liquid Reagent |
| Measurement of total chlorine, provides quicker results and clearer colors than tablets | |
| Each | |
| 1 oz. |