missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Doxycycline monohydrate
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
General Description
- Doxycycline monohydrate is a semi-synthetic tetracycline antibiotic with broad-spectrum activity
- It is a tetracycline with the 5beta-hydrogen group replaced by a hydroxy group
- Doxycycline monohydrate acts by binding the 30S ribosomal subunit and preventing polypeptide formation
- Doxycycline monohydrate may also act on the 50S ribosomal subunit, is a known inhibitor of collagenases, and has anti-inflammatory properties
Application
- Doxycycline can be used for study of cancers, infections, and antibiotic resistance in laboratory settings
- Doxycycline can be used experimentally to control TET genes and induce expression of genes of interest in vitro
Specifications
Specifications
| Type | Doxycycline Monohydrate |
| Physical Form | Powder |
| Melting Point | 167°C to 168°C |
| Color | Yellow |
| Quantity | 25 g |
| Merck Index | 14,3440 |
| Solubility Information | Soluble in ethanol or methanol; Sparingly soluble in water;Soluble in dilute acids,alkali hydroxides and dimethyl sulfoxide. |
| Formula Weight | 462.46 (444.44 Anhydrous) |
| Chemical Name or Material | Doxycycline monohydrate |
Safety and Handling
| Hazard Category | H361d |
| Hazard Statement | GHS H Statement H302 Harmful if swallowed. |
| Precautionary Statement | P201-P202-P281-P308+P313-P501c |
| TSCA | No |
| Recommended Storage | Keep cold |
RUO – Research Use Only