missing translation for 'onlineSavingsMsg'
Learn More

Doxycycline monohydrate

Catalog No. AAJ6380506
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6380506 5 g
AAJ6380514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6380506 Supplier Thermo Scientific Chemicals Supplier No. J6380506
Only null left

Doxycycline monohydrate, CAS # 17086-28-1, is the hydrated form of the tetracycline class antibiotic doxycycline. | CAS: 17086-28-1 | C22H26N2O9 | 462.455 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

General Description

  • Doxycycline monohydrate is a semi-synthetic tetracycline antibiotic with broad-spectrum activity
  • It is a tetracycline with the 5beta-hydrogen group replaced by a hydroxy group
  • Doxycycline monohydrate acts by binding the 30S ribosomal subunit and preventing polypeptide formation
  • Doxycycline monohydrate may also act on the 50S ribosomal subunit, is a known inhibitor of collagenases, and has anti-inflammatory properties

Application

  • Doxycycline can be used for study of cancers, infections, and antibiotic resistance in laboratory settings
  • Doxycycline can be used experimentally to control TET genes and induce expression of genes of interest in vitro

Chemical Identifiers

CAS 17086-28-1
Molecular Formula C22H26N2O9
Molecular Weight (g/mol) 462.455
MDL Number MFCD02682958
InChI Key FZKWRPSUNUOXKJ-CVHRZJFOSA-N
PubChem CID 54684461
IUPAC Name (4S,4aR,5S,5aR,6R,12aR)-4-(dimethylamino)-1,5,10,11,12a-pentahydroxy-6-methyl-3,12-dioxo-4a,5,5a,6-tetrahydro-4H-tetracene-2-carboxamide;hydrate
SMILES CC1C2C(C3C(C(=O)C(=C(C3(C(=O)C2=C(C4=C1C=CC=C4O)O)O)O)C(=O)N)N(C)C)O.O

Specifications

Type Doxycycline Monohydrate
Physical Form Powder
Melting Point 167°C to 168°C
Color Yellow
Quantity 5 g
Merck Index 14,3440
Solubility Information Soluble in ethanol or methanol; Sparingly soluble in water;Soluble in dilute acids,alkali hydroxides and dimethyl sulfoxide.
Formula Weight 462.46 (444.44 Anhydrous)
Chemical Name or Material Doxycycline monohydrate
Hazard Category H361d
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P201-P202-P281-P308+P313-P501c
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only