missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Doxycycline monohydrate, 97%
Chemical Identifiers
| CAS | 17086-28-1 |
|---|---|
| Molecular Formula | C22H24N2O8·H2O |
| Molecular Weight (g/mol) | 462.45 |
| InChI Key | FZKWRPSUNUOXKJ-CVHRZJFOSA-N |
| Synonym | doxycycline monohydrate, doxycycline hydrate, adoxa, doxycycline hcl, 4-dimethylamino-1,4,4a,5,5a,6,11,12a-octahydro-3,5,10,12,12a-pentahydroxy-6-methyl-1,11-dioxo-2-naphthacenecarboxamide monohydrate, adoxa pak, 4s,4ar,5s,5ar,6r,12as-4-dimethylamino-3,5,10,12,12a-pentahydroxy-6-methyl-1,11-dioxo-1,4,4a,5,5a,6,11,12a-octahydrotetracene-2-carboxamide hydrate, vibramycin-d, doxycycline usp, monodox tn |
| PubChem CID | 54684461 |
| IUPAC Name | (4S,4aR,5S,5aR,6R,12aR)-4-(dimethylamino)-1,5,10,11,12a-pentahydroxy-6-methyl-3,12-dioxo-4a,5,5a,6-tetrahydro-4H-tetracene-2-carboxamide;hydrate |
| SMILES | CC1C2C(C3C(C(=O)C(=C(C3(C(=O)C2=C(C4=C1C=CC=C4O)O)O)O)C(=O)N)N(C)C)O.O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AC458890050
|
Thermo Scientific™
458890050 |
5g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC458890250
|
Thermo Scientific™
458890250 |
25g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 17086-28-1 | |
| 462.45 | |
| doxycycline monohydrate, doxycycline hydrate, adoxa, doxycycline hcl, 4-dimethylamino-1,4,4a,5,5a,6,11,12a-octahydro-3,5,10,12,12a-pentahydroxy-6-methyl-1,11-dioxo-2-naphthacenecarboxamide monohydrate, adoxa pak, 4s,4ar,5s,5ar,6r,12as-4-dimethylamino-3,5,10,12,12a-pentahydroxy-6-methyl-1,11-dioxo-1,4,4a,5,5a,6,11,12a-octahydrotetracene-2-carboxamide hydrate, vibramycin-d, doxycycline usp, monodox tn | |
| (4S,4aR,5S,5aR,6R,12aR)-4-(dimethylamino)-1,5,10,11,12a-pentahydroxy-6-methyl-3,12-dioxo-4a,5,5a,6-tetrahydro-4H-tetracene-2-carboxamide;hydrate |
| C22H24N2O8·H2O | |
| FZKWRPSUNUOXKJ-CVHRZJFOSA-N | |
| 54684461 | |
| CC1C2C(C3C(C(=O)C(=C(C3(C(=O)C2=C(C4=C1C=CC=C4O)O)O)O)C(=O)N)N(C)C)O.O |
Specifications
| 17086-28-1 | |
| 96% min. (HPLC) | |
| 5g | |
| FZKWRPSUNUOXKJ-CVHRZJFOSA-N | |
| (4S,4aR,5S,5aR,6R,12aR)-4-(dimethylamino)-1,5,10,11,12a-pentahydroxy-6-methyl-3,12-dioxo-4a,5,5a,6-tetrahydro-4H-tetracene-2-carboxamide;hydrate | |
| 54684461 | |
| 97% |
| Authentic | |
| C22H24N2O8·H2O | |
| doxycycline monohydrate, doxycycline hydrate, adoxa, doxycycline hcl, 4-dimethylamino-1,4,4a,5,5a,6,11,12a-octahydro-3,5,10,12,12a-pentahydroxy-6-methyl-1,11-dioxo-2-naphthacenecarboxamide monohydrate, adoxa pak, 4s,4ar,5s,5ar,6r,12as-4-dimethylamino-3,5,10,12,12a-pentahydroxy-6-methyl-1,11-dioxo-1,4,4a,5,5a,6,11,12a-octahydrotetracene-2-carboxamide hydrate, vibramycin-d, doxycycline usp, monodox tn | |
| CC1C2C(C3C(C(=O)C(=C(C3(C(=O)C2=C(C4=C1C=CC=C4O)O)O)O)C(=O)N)N(C)C)O.O | |
| 462.45 | |
| 462.45 | |
| Doxycycline monohydrate |
Safety and Handling
GHS H Statement
Suspected of damaging the unborn child.
GHS P Statement
Obtain special instructions before use.
Use personal protective equipment as required.
IF exposed or concerned: Get medical advice/attention.
GHS Signal Word: Warning
RUO â Research Use Only