missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Doxycycline monohydrate, 97%

Catalog No. AC458890250
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC458890250 25 g
AC458890050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC458890250 Supplier Thermo Scientific Chemicals Supplier No. 458890250
Only null left

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 17086-28-1
Molecular Formula C22H24N2O8·H2O
Molecular Weight (g/mol) 462.45
InChI Key FZKWRPSUNUOXKJ-CVHRZJFOSA-N
Synonym doxycycline monohydrate, doxycycline hydrate, adoxa, doxycycline hcl, 4-dimethylamino-1,4,4a,5,5a,6,11,12a-octahydro-3,5,10,12,12a-pentahydroxy-6-methyl-1,11-dioxo-2-naphthacenecarboxamide monohydrate, adoxa pak, 4s,4ar,5s,5ar,6r,12as-4-dimethylamino-3,5,10,12,12a-pentahydroxy-6-methyl-1,11-dioxo-1,4,4a,5,5a,6,11,12a-octahydrotetracene-2-carboxamide hydrate, vibramycin-d, doxycycline usp, monodox tn
PubChem CID 54684461
IUPAC Name (4S,4aR,5S,5aR,6R,12aR)-4-(dimethylamino)-1,5,10,11,12a-pentahydroxy-6-methyl-3,12-dioxo-4a,5,5a,6-tetrahydro-4H-tetracene-2-carboxamide;hydrate
SMILES CC1C2C(C3C(C(=O)C(=C(C3(C(=O)C2=C(C4=C1C=CC=C4O)O)O)O)C(=O)N)N(C)C)O.O

Specifications

Melting Point 167.0°C to 168.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Quantity 25 g
Formula Weight 462.45
Percent Purity 97%
Chemical Name or Material Doxycycline monohydrate
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Suspected of damaging the unborn child.
Precautionary Statement GHS P Statement
Obtain special instructions before use.
Use personal protective equipment as required.
IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only