missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Doxycycline monohydrate, 97%
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 167.0°C to 168.0°C |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 96% min. (HPLC) |
| Quantity | 5 g |
| Formula Weight | 462.45 |
| Percent Purity | 97% |
| Chemical Name or Material | Doxycycline monohydrate |
Safety and Handling
| Hazard Category | GHS Signal Word: Warning |
| Hazard Statement | GHS H Statement Suspected of damaging the unborn child. |
| Precautionary Statement | GHS P Statement Obtain special instructions before use. Use personal protective equipment as required. IF exposed or concerned: Get medical advice/attention. |
RUO – Research Use Only