Learn More
Doxycycline hyclate
Doxycycline hyclate, Doryx, CAS # 24390-14-5, is the hyclate salt form of the broad-spectrum antibiotic known as doxycycline. | CAS: 24390-14-5 | C26H41ClN2O12 | 609.066 g/mol
$201.95 - $649.38
Chemical Identifiers
| CAS | 24390-14-5 |
|---|---|
| Molecular Formula | C26H41ClN2O12 |
| Molecular Weight (g/mol) | 609.066 |
| MDL Number | MFCD07357237 |
| InChI Key | ISZOFWGVNMFENH-YDLUHMIOSA-N |
| PubChem CID | 131676157 |
| IUPAC Name | (4S,4aR,5S,5aR,6R,12aR)-4-(dimethylamino)-1,5,10,11,12a-pentahydroxy-6-methyl-3,12-dioxo-4a,5,5a,6-tetrahydro-4H-tetracene-2-carboxamide;ethanol;dihydrate;hydrochloride |
| SMILES | CCO.CCO.CC1C2C(C3C(C(=O)C(=C(C3(C(=O)C2=C(C4=C1C=CC=C4O)O)O)O)C(=O)N)N(C)C)O.O.O.Cl |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
AAJ6057914
|
N/A | 25 g |
Each for $201.95
|
|
|||||
|
AAJ6057922
|
N/A | 100 g |
Each for $649.38
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
General description
• Doxycycline hyclate is a broad-spectrum bacteriostatic antibiotic of the tetracycline class
• Doxycycline inhibits bacterial growth by preventing polypeptide formation via inhibition of at least one ribosomal subunit
• Known to reversibly bind the 30S ribosomal subunit and inhibit formation of the aminoacyl-tRNA/mRNA-ribosome complex
• Inhibits matrix metalloproteinases in many microorganisms
Applications
• Effective against the bacteria that cause Lyme disease, chlamydia, rickettsia and mycoplasma; suitable for antibiotic resistance testing and study of cellular mechanisms
• Intended for laboratory use only
Chemical Identifiers
| 24390-14-5 | |
| 609.066 | |
| ISZOFWGVNMFENH-YDLUHMIOSA-N | |
| (4S,4aR,5S,5aR,6R,12aR)-4-(dimethylamino)-1,5,10,11,12a-pentahydroxy-6-methyl-3,12-dioxo-4a,5,5a,6-tetrahydro-4H-tetracene-2-carboxamide;ethanol;dihydrate;hydrochloride |
| C26H41ClN2O12 | |
| MFCD07357237 | |
| 131676157 | |
| CCO.CCO.CC1C2C(C3C(C(=O)C(=C(C3(C(=O)C2=C(C4=C1C=CC=C4O)O)O)O)C(=O)N)N(C)C)O.O.O.Cl |
Specifications
| Doxycycline Hyclate | |
| 24390-14-5 | |
| C26H41ClN2O12 | |
| 14,3440 | |
| Soluble in water to 50mg/ml; May require warming;Soluble in water,methanol; Sparingly soluble in ethanol; Insoluble in chloroform and ether. | |
| CCO.CCO.CC1C2C(C3C(C(=O)C(=C(C3(C(=O)C2=C(C4=C1C=CC=C4O)O)O)O)C(=O)N)N(C)C)O.O.O.Cl | |
| 609.066 | |
| 512.94 (503.94 Anhydrous) |
| Powder | |
| Yellow | |
| MFCD07357237 | |
| Hydramycin; Doxycycline hydrochloride hemiethanolate hemihydrate | |
| ISZOFWGVNMFENH-YDLUHMIOSA-N | |
| (4S,4aR,5S,5aR,6R,12aR)-4-(dimethylamino)-1,5,10,11,12a-pentahydroxy-6-methyl-3,12-dioxo-4a,5,5a,6-tetrahydro-4H-tetracene-2-carboxamide;ethanol;dihydrate;hydrochloride | |
| 131676157 | |
| Doxycycline hyclate |
Safety and Handling
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P201-P202-P261-P264b-P270-P271-P280i-P281-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P330-P332+P313-P362-P501c
H302-H315-H319-H335-H361fd
RTECSNumber : QI8925000
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only