missing translation for 'onlineSavingsMsg'
Learn More

Doxycycline Hyclate 97.0+%, TCI America™

Supplier:  TCI America D41165G

 View more versions of this product

Catalog No. D41165G

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Doxycycline Hyclate
Yellow
MFCD07357237
doryx, doxycycline hyclate, doxycycline, hydrochloride, hemiethanolate, hemihydrate;
CCO.CCO.CC1C2C(C3C(C(=O)C(=C(C3(C(=O)C2=C(C4=C1C=CC=C4O)O)O)O)C(=O)N)N(C)C)O.O.O.Cl
609.066
503.93
Crystalline Powder
24390-14-5
C26H41ClN2O12
5 g
ISZOFWGVNMFENH-YDLUHMIOSA-N
(4S,4aR,5S,5aR,6R,12aR)-4-(dimethylamino)-1,5,10,11,12a-pentahydroxy-6-methyl-3,12-dioxo-4a,5,5a,6-tetrahydro-4H-tetracene-2-carboxamide;ethanol;dihydrate;hydrochloride
131676157
≥97.0% (HPLC)

Chemical Identifiers

24390-14-5
609.066
ISZOFWGVNMFENH-YDLUHMIOSA-N
131676157
CCO.CCO.CC1C2C(C3C(C(=O)C(=C(C3(C(=O)C2=C(C4=C1C=CC=C4O)O)O)O)C(=O)N)N(C)C)O.O.O.Cl
C26H41ClN2O12
MFCD07357237
doryx, doxycycline hyclate, doxycycline, hydrochloride, hemiethanolate, hemihydrate;
(4S,4aR,5S,5aR,6R,12aR)-4-(dimethylamino)-1,5,10,11,12a-pentahydroxy-6-methyl-3,12-dioxo-4a,5,5a,6-tetrahydro-4H-tetracene-2-carboxamide;ethanol;dihydrate;hydrochloride
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions