missing translation for 'onlineSavingsMsg'
Learn More

Doxorubicin Hydrochloride 95.0+%, TCI America™

Catalog No. d419325mg
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
100 mg
25 mg
Catalog No. Quantity
D419325MG 25 mg
D4193100MG 100 mg
2 options

Catalog No. D419325MG

Supplier: TCI America D419325MG

Only null left

Chemical Identifiers

CAS 25316-40-9
Molecular Formula C27H30ClNO11
Molecular Weight (g/mol) 579.98
MDL Number MFCD00077757,MFCD00077757,MFCD00941448
InChI Key MWWSFMDVAYGXBV-FGBJBKNOSA-N
Synonym doxorubicin hydrochloride, adriamycin, doxorubicin hcl, adriacin, adriblastina, rubex, adriamycin hydrochloride, adriblastin, adriamycin pfs
PubChem CID 129626538
IUPAC Name hydrogen (8S,10S)-10-[(4-amino-5-hydroxy-6-methyloxan-2-yl)oxy]-6,8,11-trihydroxy-8-(2-hydroxyacetyl)-1-methoxy-5,7,8,9,10,12-hexahydrotetracene-5,12-dione chloride
SMILES [H+].[Cl-].COC1=CC=CC2=C1C(=O)C1=C(O)C3=C(C[C@](O)(C[C@@H]3OC3CC(N)C(O)C(C)O3)C(=O)CO)C(O)=C1C2=O

Specifications

Quantity 25 mg
Formula Weight 579.98
Percent Purity ≥95.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Doxorubicin Hydrochloride
RTECSNumber QI9295900
TSCA No
Recommended Storage Refrigerator