missing translation for 'onlineSavingsMsg'
Learn More

Doxofylline

Catalog No. AAJ6057506
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6057506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ6057506 Supplier Thermo Scientific Chemicals Supplier No. J6057506

missing translation for 'validMainframeMsg'

CAS: 69975-86-6 | C11H14N4O4 | 266.257 g/mol

Xanthine bronchodilator that inhibits phosphodiesterase activity

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 69975-86-6
Molecular Formula C11H14N4O4
Molecular Weight (g/mol) 266.257
MDL Number MFCD00865218
InChI Key HWXIGFIVGWUZAO-UHFFFAOYSA-N
Synonym doxofylline, doxophylline, ansimar, dioxyfilline, maxivent, ventax, unii-mpm23gmo7z, doxofylline usan:inn, doxofilina inn-spanish, doxofyllinum inn-latin
PubChem CID 50942
IUPAC Name 7-(1,3-dioxolan-2-ylmethyl)-1,3-dimethylpurine-2,6-dione
SMILES CN1C2=C(C(=O)N(C1=O)C)N(C=N2)CC3OCCO3

Specifications

Melting Point 144°C to 146°C
Color White
Quantity 5 g
Merck Index 14,3438
Formula Weight 266.25
Physical Form Crystalline Powder
Chemical Name or Material Doxofylline
Hazard Category H302
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 274-239-6
RTECSNumber XH5135000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only