missing translation for 'onlineSavingsMsg'
Learn More

Doxofylline 98.0+%, TCI America™

Catalog No. p-7128209
Change view
Click to view available options
Quantity:
1 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D43021G 1 g
D430225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D43021G Supplier TCI America Supplier No. D43021G
Only null left

Chemical Identifiers

CAS 69975-86-6
Molecular Formula C11H14N4O4
Molecular Weight (g/mol) 266.257
MDL Number MFCD00865218
InChI Key HWXIGFIVGWUZAO-UHFFFAOYSA-N
Synonym doxofylline, doxophylline, ansimar, dioxyfilline, maxivent, ventax, unii-mpm23gmo7z, doxofylline usan:inn, doxofilina inn-spanish, doxofyllinum inn-latin
PubChem CID 50942
IUPAC Name 7-(1,3-dioxolan-2-ylmethyl)-1,3-dimethylpurine-2,6-dione
SMILES CN1C2=C(C(=O)N(C1=O)C)N(C=N2)CC3OCCO3

Specifications

Melting Point 144°C
Color White
Quantity 1 g
Formula Weight 266.26
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Doxofylline
RTECSNumber XH5135000
TSCA No