missing translation for 'onlineSavingsMsg'
Learn More

Doxepin Hydrochloride (mixture of isomers) 98.0+%, TCI America™

Catalog No. D46265G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D46265G 5 g
D46261G 1 g
D462625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D46265G Supplier TCI America Supplier No. D46265G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1229-29-4
Molecular Formula C19H22ClNO
Molecular Weight (g/mol) 315.841
MDL Number MFCD00079135
InChI Key MHNSPTUQQIYJOT-SJDTYFKWSA-N
Synonym doxepin hydrochloride, novoxapin, aponal, toruan, curatin, sinequan, zonalon, adapin, doxepine hydrochloride, novoxapin hydrochloride
PubChem CID 6419921
IUPAC Name (3E)-3-(6H-benzo[c][1]benzoxepin-11-ylidene)-N,N-dimethylpropan-1-amine;hydrochloride
SMILES CN(C)CCC=C1C2=CC=CC=C2COC3=CC=CC=C31.Cl

Specifications

Melting Point 188°C
Color White-Yellow
Quantity 5 g
UN Number 2811
Formula Weight 315.84
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Doxepin Hydrochloride (mixture of isomers)
RTECSNumber HQ4375000
TSCA No