missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ Dowex™ 50WX8 Hydrogen Form, Strongly Acidic, 50-100 mesh

Catalog No. 111015244
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015244 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015244 Supplier MilliporeSigma™ Supelco™ Supplier No. 44509100G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Dowex™ 50WX8 hydrogen form may be used in separation medium for TLC, paper chromatography and LPLC.

Dowex™ 50WX8 hydrogen form is used as stripper resin and can be regenerated by 0.25N sulphuric acid when exhausted. Dowex™ 50WX8 hydrogen form is used as catalyst. It is used in deionizing supernatant fraction samples for paper chromatography.

Specifications

Particle Size 50/100 Mesh
Technique Chromatography
Quantity 100 g
Product Line DOWEX
Material Styrene-divinylbenzene
Capacity (Metric) 1.1 meq/mL by wetted bed volume
MDL Number MFCD00132726
CAS 69011-20-7

DOWEX is a registered trademark of The Dow Chemical Company or an affiliated company of Dow