missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ Dowex™ 50WX8 Hydrogen Form, 100-200 mesh

Catalog No. 111015224
Change view
Click to view available options
Quantity:
2.5 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015224 2.5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015224 Supplier MilliporeSigma™ Supelco™ Supplier No. 2175062.5KG

This resin has not been specially processed nor cleaned. It is suggested that it be sujbected to a suitable preliminary elution and wash.

Dowex™ 50WX8 hydrogen form is used as stripper resin and can be regenerated by 0.25N sulphuric acid when exhausted. Dowex™ 50WX8 hydrogen form is used as catalyst. It is used in deionizing supernatant fraction samples for paper chromatography.

Specifications

Particle Size 100/200 Mesh
Packaging Poly Bottle
Quantity 2.5 kg
Product Line DOWEX
Capacity (Metric) 1.7 meq/mL by wetted bed volume
MDL Number MFCD00132726
CAS 69011-20-7

DOWEX is a registered trademark of The Dow Chemical Company or an affiliated company of Dow