missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dowex™ 50WX4 100-200 (H)
Description
Dowex™ 50WX4 100-200 (H) is used for the conversion of D,L-glyceraldehyde 3-phosphate diethyl acetal (barium salt) on treatment with boiling water to free aldehyde D,L-glyceraldehyde 3-phosphate. It is also used to investigate the effect of particle size on drug loading and release profiles.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Dowex|r 50WX4 100-200 (H) is used for the conversion of D,L-glyceraldehyde 3-phosphate diethyl acetal (barium salt) on treatment with boiling water to free aldehyde D,L-glyceraldehyde 3-phosphate. It is also used to investigate the effect of particle size on drug loading and release profiles.
Solubility
Insoluble in water.
Notes
Incompatible with oxidizing agents.
Dowex|r 50WX4 100-200 (H) is used for the conversion of D,L-glyceraldehyde 3-phosphate diethyl acetal (barium salt) on treatment with boiling water to free aldehyde D,L-glyceraldehyde 3-phosphate. It is also used to investigate the effect of particle size on drug loading and release profiles.
Solubility
Insoluble in water.
Notes
Incompatible with oxidizing agents.
Specifications
Specifications
| Quantity | 50 g |
| Solubility Information | Insoluble in water. |
| Chemical Name or Material | Dowex™ 50WX4 100-200 (H) |
Safety and Handling
| Hazard Category | H318 |
| Precautionary Statement | P280-P305+P351+P338-P310 |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only