missing translation for 'onlineSavingsMsg'
Learn More

Dowex™ 1X8 100-200 (Cl)

CAS: 12627-85-9 | C29H34ClN | 432.05 g/mol

Supplier:  Thermo Scientific Chemicals L142570B

 View more versions of this product

Catalog No. AAL142570B

Only null left
Explore available promotions

Description

Description

Dowex™ 1X8 100-200 (Cl) is used as an absorbent in solid phase extraction. It is used in anion-exchange chromatography for separation. It also finds application in fine chemical processing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dowex|r 1X8 100-200 (Cl) is used as an absorbent in solid phase extraction. It is used in anion-exchange chromatography for separation. It also finds application in fine chemical processing.

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents.
Specifications

Specifications

Dowex™ 1X8 100-200 (Cl)
Amine-like
MFCD00132718
dowex 1x2 50-100 cl, dowex 1x8 50-100 cl
BBQMUEOYPPPODD-UHFFFAOYSA-M
1,4-diethenylbenzene 4-ethenyl-N,N,N-trimethylanilinium ethenylbenzene chloride
16212807
12627-85-9
C29H34ClN
1000 g
Insoluble in water.
*
432.05

Chemical Identifiers

12627-85-9
432.05
BBQMUEOYPPPODD-UHFFFAOYSA-M
16212807
*
C29H34ClN
MFCD00132718
dowex 1x2 50-100 cl, dowex 1x8 50-100 cl
1,4-diethenylbenzene 4-ethenyl-N,N,N-trimethylanilinium ethenylbenzene chloride
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only