missing translation for 'onlineSavingsMsg'
Learn More

AmberChrom™ 1X8 100-200 (Cl)

Catalog No. AAL1425730
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1425730 250 g
AAL1425718 50 g
AAL142570B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1425730 Supplier Thermo Scientific Chemicals Supplier No. L1425730

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 12627-85-9 | C29H34ClN | 432.05 g/mol

AmberChrom™ 1X8 100-200 (Cl) is used as an absorbent in solid phase extraction. It is used in anion-exchange chromatography for separation. It also finds application in fine chemical processing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
AmberChrom 1X8 100-200 (Cl) is used as an absorbent in solid phase extraction. It is used in anion-exchange chromatography for separation. It also finds application in fine chemical processing.

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 12627-85-9
Molecular Formula C29H34ClN
Molecular Weight (g/mol) 432.05
MDL Number MFCD00132718
InChI Key BBQMUEOYPPPODD-UHFFFAOYSA-M
Synonym dowex 1x2 50-100 cl, dowex 1x8 50-100 cl
PubChem CID 16212807
IUPAC Name 1,4-diethenylbenzene 4-ethenyl-N,N,N-trimethylanilinium ethenylbenzene chloride
SMILES *

Specifications

Odor Amine-like
Quantity 250 g
Solubility Information Insoluble in water.
Chemical Name or Material AmberChrom 1X8 100-200 (Cl)
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only