missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dowex™ 1X2, 200-400 mesh, ion-exchange resin
CAS: 9085-42-1 | C29H34ClN | 432.05 g/mol
$464.48 - $464.48
Chemical Identifiers
| CAS | 9085-42-1 |
|---|---|
| Molecular Formula | C29H34ClN |
| Molecular Weight (g/mol) | 432.05 |
| MDL Number | MFCD00132715 |
| InChI Key | BBQMUEOYPPPODD-UHFFFAOYSA-M |
| Synonym | dowex 1x2 50-100 cl, dowex 1x8 50-100 cl |
| PubChem CID | 16212807 |
| SMILES | [Cl-].C=CC1=CC=CC=C1.C=CC1=CC=C(C=C)C=C1.C[N+](C)(C)C1=CC=C(C=C)C=C1 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC202975000
|
Thermo Scientific Chemicals
202975000 |
500 g | Plastic bottle |
Each for $464.48
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Ion exchange resinChemical Identifiers
| 9085-42-1 | |
| 432.05 | |
| BBQMUEOYPPPODD-UHFFFAOYSA-M | |
| 16212807 |
| C29H34ClN | |
| MFCD00132715 | |
| dowex 1x2 50-100 cl, dowex 1x8 50-100 cl | |
| [Cl-].C=CC1=CC=CC=C1.C=CC1=CC=C(C=C)C=C1.C[N+](C)(C)C1=CC=C(C=C)C=C1 |
Specifications
| Beads | |
| Particle size analysis: on 70 Mesh (cumulative): <15%, through 270 Mesh: <15% | |
| wet volume capacity, Cl form (meq/mL):> 0.6 | |
| MFCD00132715 | |
| 01,511; 04,267; 06,302; 07,182; 08,263 | |
| 500 g | |
| Solubility in water: insoluble. | |
| [Cl-].C=CC1=CC=CC=C1.C=CC1=CC=C(C=C)C=C1.C[N+](C)(C)C1=CC=C(C=C)C=C1 | |
| 432.05 | |
| Dowex™ 1X2 |
| 9085-42-1 | |
| Plastic bottle | |
| C29H34ClN | |
| Cl form: 70 to 80% | |
| 10, 2182 | |
| dowex 1x2 50-100 cl, dowex 1x8 50-100 cl | |
| BBQMUEOYPPPODD-UHFFFAOYSA-M | |
| 1,4-bis(ethenyl)benzene;(4-ethenylphenyl)-trimethylazanium;styrene;chloride | |
| 16212807 |
RUO – Research Use Only