missing translation for 'onlineSavingsMsg'
Learn More

Dowex™ 1X2 100-200 (Cl)

CAS: 9085-42-1 | C29H34ClN | 432.05 g/mol

Supplier:  Thermo Scientific Chemicals L1425418

 View more versions of this product

Catalog No. AAL1425418

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Dowex™ 1X2 100-200 (Cl)
Amine-like
MFCD00132715
dowex 1x2 50-100 cl, dowex 1x8 50-100 cl
[Cl-].C=CC1=CC=CC=C1.C=CC1=CC=C(C=C)C=C1.C[N+](C)(C)C1=CC=C(C=C)C=C1
432.05
9085-42-1
C29H34ClN
50 g
BBQMUEOYPPPODD-UHFFFAOYSA-M
1,4-bis(ethenyl)benzene;(4-ethenylphenyl)-trimethylazanium;styrene;chloride
16212807

Chemical Identifiers

9085-42-1
432.05
BBQMUEOYPPPODD-UHFFFAOYSA-M
16212807
[Cl-].C=CC1=CC=CC=C1.C=CC1=CC=C(C=C)C=C1.C[N+](C)(C)C1=CC=C(C=C)C=C1
C29H34ClN
MFCD00132715
dowex 1x2 50-100 cl, dowex 1x8 50-100 cl
1,4-bis(ethenyl)benzene;(4-ethenylphenyl)-trimethylazanium;styrene;chloride
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only