missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dowex™ 1X2 100-200 (Cl)
CAS: 9085-42-1 | C29H34ClN | 432.05 g/mol
Supplier: Thermo Scientific Chemicals L142540B
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Dowex™ 1X2 100-200 (Cl) | |
| Amine-like | |
| MFCD00132715 | |
| dowex 1x2 50-100 cl, dowex 1x8 50-100 cl | |
| [Cl-].C=CC1=CC=CC=C1.C=CC1=CC=C(C=C)C=C1.C[N+](C)(C)C1=CC=C(C=C)C=C1 | |
| 432.05 |
| 9085-42-1 | |
| C29H34ClN | |
| 1000 g | |
| BBQMUEOYPPPODD-UHFFFAOYSA-M | |
| 1,4-bis(ethenyl)benzene;(4-ethenylphenyl)-trimethylazanium;styrene;chloride | |
| 16212807 |
Chemical Identifiers
| 9085-42-1 | |
| 432.05 | |
| BBQMUEOYPPPODD-UHFFFAOYSA-M | |
| 16212807 | |
| [Cl-].C=CC1=CC=CC=C1.C=CC1=CC=C(C=C)C=C1.C[N+](C)(C)C1=CC=C(C=C)C=C1 |
| C29H34ClN | |
| MFCD00132715 | |
| dowex 1x2 50-100 cl, dowex 1x8 50-100 cl | |
| 1,4-bis(ethenyl)benzene;(4-ethenylphenyl)-trimethylazanium;styrene;chloride |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only