missing translation for 'onlineSavingsMsg'
Learn More

Dowex™ 1X2 100-200 (Cl)

Catalog No. AAL1425430
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1425430 250 g
AAL1425418 50 g
AAL142540B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1425430 Supplier Thermo Scientific Chemicals Supplier No. L1425430

missing translation for 'validMainframeMsg'

CAS: 9085-42-1 | C29H34ClN | 432.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 9085-42-1
Molecular Formula C29H34ClN
Molecular Weight (g/mol) 432.05
MDL Number MFCD00132715
InChI Key BBQMUEOYPPPODD-UHFFFAOYSA-M
Synonym dowex 1x2 50-100 cl, dowex 1x8 50-100 cl
PubChem CID 16212807
IUPAC Name 1,4-bis(ethenyl)benzene;(4-ethenylphenyl)-trimethylazanium;styrene;chloride
SMILES [Cl-].C=CC1=CC=CC=C1.C=CC1=CC=C(C=C)C=C1.C[N+](C)(C)C1=CC=C(C=C)C=C1

Specifications

Odor Amine-like
Quantity 250 g
Chemical Name or Material Dowex™ 1X2 100-200 (Cl)
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only