Electron Microscopy Sciences Conductive Double Sided Carbon Tape, 12 mm
Catalog No. 5028580
Can be used to mount particles 15-20 microns. Isolate unwoved cloth. Adhesive power/25mm: 1000 gf. Conductive resistivity: 50ohm/sq.Inch Thickness: 0.16 mm Pad: 0.07 mm Adhesive: 0.045 mm x 2
Catalog No.
5028580
Supplier
Electron Microscopy SciencesSupplier No.
7781712
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Can be used to mount particles 15-20 microns. Isolate unwoved cloth. Adhesive power/25mm: 1000 gf. Conductive resistivity: 50ohm/sq.Inch Thickness: 0.16 mm Pad: 0.07 mm Adhesive: 0.045 mm x 2