missing translation for 'onlineSavingsMsg'
Learn More

Synthware™ Double Oblique Bore Glass Stopcock
GREENER_CHOICE

Catalog No. 31502044
Change view
Click to view available options
Plug Bore:
4 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Plug Bore
31502044 4 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 31502044 Supplier Synthware™ Supplier No. S351656

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Double Oblique Bore Stopcock has diagonal, parallel bores to allow connection of single stem to both stems on the opposite side. Hollow glass plug option is precision ground with corresponding numbers on the barrel and the plug

Specifications

Type Double Oblique Stopcock
Plug Bore 4 mm
Diameter (Metric) 10 mm
Material Borosilicate Glass
For Use With (Application) Organic Synthesis
Plug Material Glass
Connectors Ground Glass