missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Synthware™ Double Oblique Bore PTFE Stopcocks

Specifications
Specifications
| Type | Double Oblique Stopcock |
| Plug Bore | 4 mm |
| Diameter (Metric) | 10 mm |
| Material | Borosilicate Glass |
| For Use With (Application) | Organic Synthesis |
| Plug Material | PTFE |
| Connectors | Hose |