missing translation for 'onlineSavingsMsg'
Learn More

National Safety Apparel™ 40 cal Replacement Fan for Crossvent Hood

Catalog No. p-7220312
Change view
Click to view available options
Style:
Left Side
Right Side
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Style
17001689 Left Side
17001691 Right Side
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 17001689 Supplier National Safety Apparel™ Supplier No. FANUQUQLS
Only null left

Crossvent hood replacement fan

Specifications

Size Medium
Material Plastic
Color Navy
Seam Type Stitched
For Use With (Equipment) Arc Flash Hoods
Includes Left Side Fan
Style Left Side