missing translation for 'onlineSavingsMsg'
Learn More

National Safety Apparel™ 40 cal Replacement Fan for Crossvent Hood

Catalog No. 17001691
Encompass
Change view
Click to view available options
Style:
Left Side
Right Side
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Style
17001691 Right Side
17001689 Left Side
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 17001691 Supplier National Safety Apparel™ Supplier No. FANUQUQRS

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Crossvent hood replacement fan

Specifications

Size Medium
Material Plastic
Color Navy
Seam Type Stitched
For Use With (Equipment) Arc Flash Hoods
Includes Right Side Fan
Style Right Side