Double Deck Tube Rocker holds up to 30 test tubes and silently rocks them at a fixed rate of 25 RPM for gentle mixing Blood Tube Rocker perfect for busy hematology departmentsNow with new integrated "pause" button which allows you to stop the "rocking" temporarily in order to retrieve tubes without burning up the motor25 RPM rocking motion preserves integrity of fragile blood cellsProtects samples from being hemolyzed or traumatizedGreat for mixing control samples before usePerfect for slide preparation areas to use before making a smear when Diff is indicatedThe reversible non-slip pad is autoclavable and will accommodate larger and hard to mix tubesHolds 30 tubes up to 120mm longCompact size uses minimal bench spaceOverall Dimensions: 13.7"W x 4.7"D x 7"H