Learn More
United Scientific™ Double Convex Lens
Clear Lens for consistent optics demonstrations.
Supplier:  United Scientific™ LCV508
 View more versions of this product
Catalog No. S01322
Only null left
Alert:
Prop 65
Description
Description
  • Made from plate glass.
  • Features carefully ground and polished optical surfaces.
  • Cleanly ground edges.
  • Double Convex.
Specifications
Specifications
1 Ea.
2 in.
200 mm
50 mm
7.9 in.
Convex Lens
Product Suggestions
Product Suggestions