Learn More
Eisco™ Double Concave Glass Lens
Perform experiments with high quality, optically true lenses. The Double Concave Glass Lens is made of optically worked, highly polished glass with ground edges.
Supplier:  Eisco™ PH0536CCC
 View more versions of this product
Catalog No. S23673
Only null left
Description
Description
  • Spherical
  • Optically worked glass
  • Ground edges
  • Highly polished
Specifications
Specifications
Glass
150 mm
Double Concave
38 mm
Double Concave Lens
Product Suggestions
Product Suggestions