Learn More
Eisco™ Double Concave Glass Lens
Perform experiments with high quality, optically true lenses. The Double Concave Glass Lens is made of optically worked, highly polished glass with ground edges.
Supplier:  Eisco™ PH0536QCC
 View more versions of this product
Catalog No. S27395
Only null left
Description
Description
  • Spherical
  • Optically worked glass
  • Ground edges
  • Highly polished
Specifications
Chemical Identifiers
Glass
Double Concave Lens
Specifications
Glass
3 in.
250 mm
Double Concave Circle
75 mm
Educational Purposes, Top Spinning
Double Concave Lens
Product Suggestions
Product Suggestions