missing translation for 'onlineSavingsMsg'
Learn More

Brady™ DOT Vehicle Placards: OXIDIZER 5.1

Catalog No. 19111352
Change view
Click to view available options
Material:
Fiberglass (B-120)
Polycoated Tagstock (B-101)
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
19111352 Polycoated Tagstock (B-101)
19111336 Fiberglass (B-120)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19111352 Supplier Brady™ Supplier No. 63449

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Comply with DOT regulations by marking delivery vehicles carrying dangerous or hazardous materials. Brady™ DOT Vehicle Placards are available in a variety of materials.

  • Available in three different materials Pressure Sensitive Vinyl, Polycoated Tagstock and Premium Fiberglass

Specifications

Adhesive Type Non-adhesive
Color White, Blue
Finish Semi-gloss
Format Label
Material Polycoated Tagstock (B-101)
Product Type Vehicle Placard
Sterility Non-sterile
Display Text OXIDIZER 5.1
For Use With (Application) Material Handling, Shipping and Receiving Signs and Labels
Quantity 1 Ea.
Shape Diamond
Size (English) 10.75 x 10.75 in.
Thickness (English) 0.0024 in.
Dimensions (L x W) 10.75 x 10.75 in.
Length (English) 10.75 in.
Width (English) 10.75 in.
Show More Show Less