missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ DOT Vehicle Placards: OXIDIZER 5.1
Comply with DOT regulations by marking delivery vehicles carrying dangerous or hazardous materials. Brady™ DOT Vehicle Placards are available in a variety of materials.
Supplier: Brady™ 63428
Description
- Available in three different materials Pressure Sensitive Vinyl, Polycoated Tagstock and Premium Fiberglass
Specifications
| Non-adhesive | |
| Matte | |
| Fiberglass (B-120) | |
| Non-sterile | |
| Material Handling, Shipping and Receiving Signs and Labels | |
| Diamond | |
| 0.0984 in. | |
| 10.75 in. |
| White, Blue | |
| Label | |
| Vehicle Placard | |
| OXIDIZER 5.1 | |
| 1 Ea. | |
| 10.75 x 10.75 in. | |
| 10.75 x 10.75 in. |