missing translation for 'onlineSavingsMsg'
Learn More

Brady™ DOT Vehicle Placards: EXPLOSIVE 1.1E

Comply with DOT regulations by marking delivery vehicles carrying dangerous or hazardous materials. Brady™ DOT Vehicle Placards are available in a variety of materials.

$37.47 - $37.47

Specifications

Color Black, Orange
Product Type Vehicle Placard
Display Text EXPLOSIVE 1.1E
For Use With (Application) Material Handling, Shipping and Receiving Signs and Labels
Quantity 1 Ea.
View More Specs

Products 1
Catalog Number Mfr. No. Material Price Quantity  
Catalog Number Mfr. No. Material Price Quantity  
19-111-268
View Documents
Brady™
63339
Fiberglass
Each for $37.47
Only null left
 
Description

Description

  • Available in two different materials Premium Fiberglass, Pressure Sensitive Vinyl
Specifications

Specifications

Black, Orange
EXPLOSIVE 1.1E
1 Ea.
10.75 x 10.75 in.
Vehicle Placard
Material Handling, Shipping and Receiving Signs and Labels
Diamond
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'