missing translation for 'onlineSavingsMsg'
Learn More

Brady™ DOT Standard Placards: (Flammable Picto w/Blank Box) 3

Comply with DOT regulations by marking delivery vehicles carrying dangerous or hazardous materials. Brady™ DOT Standard Placards are available in a variety of materials.

$16.30 - $16.30

Specifications

Color White, Red
Product Type Vehicle Placard
Display Text Flammable pictogram with blank box, 3
For Use With (Application) Material Handling, Shipping and Receiving Signs and Labels
Quantity 1 Ea.
View More Specs

Products 1
Catalog Number Mfr. No. Material Price Quantity  
Catalog Number Mfr. No. Material Price Quantity  
19-110-997
View Documents
Brady™
60378
Vinyl (B-946)
Each for $16.30
Only null left
 
Description

Description

  • Available in three different materials Pressure Sensitive Vinyl, Polycoated Tagstock and Premium Fiberglass
Specifications

Specifications

White, Red
Flammable pictogram with blank box, 3
1 Ea.
10.75 x 10.75 in.
Vehicle Placard
Material Handling, Shipping and Receiving Signs and Labels
Diamond
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'