missing translation for 'onlineSavingsMsg'
Learn More

Accuform Signs DOT Shipping Labels

to place proper identification and classification of hazardous material during transporting and shipping

Supplier:  Accuform Signs MSL202PS2

 View more versions of this product

Catalog No. 19809835


Only null left

Description

Description

Specifications

Chemical Identifiers

Self-adhesive
250/Pk.
10.16 cm
Label
4 in.

Specifications

Self-adhesive
Paper
Flammable Gas 2
Graphic
4 in.
10.16 cm
Label
DOT Shipping Label
Shipping
250/Pk.
4 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions